Merck
  • Biomarker measurements in a coastal fish-eating population environmentally exposed to organochlorines.
Biomarker measurements in a coastal fish-eating population environmentally exposed to organochlorines.
Environmental health perspectives (2005-10-06)
Pierre Ayotte, Eric Dewailly, George H Lambert, Sherry L Perkins, Raymond Poon, Mark Feeley, Christian Larochelle, Daria Pereg
ABSTRACT
The Lower North Shore region of the St. Lawrence River is home to a fish-eating population that displays an unusually high body burden of several organochlorines, including polychlorinated biphenyls (PCBs) and dioxin-like compounds (DLCs). We measured biomarkers indicative of liver enzyme induction and investigated the relationship with organochlorine body burden in adult volunteers from this population. We determined plasma concentrations of PCBs and chlorinated pesticides by high-resolution gas chromatography (HRGC) with electron capture detection. DLC concentrations were measured by the dioxin-receptor chemically activated luciferase expression (DR-CALUX) assay and in a subset of participants, by HRGC/high-resolution mass spectrometry. We measured cotinine, d-glucaric acid, and porphyrins in morning urine samples and determined liver CYP1A2 activity in vivo using the caffeine breath test. Neither DLC concentrations as measured by the DR-CALUX nor PCB-153 concentrations, the latter representing total PCB exposure, were correlated with biomarkers of effects. Smoking (morning urinary cotinine concentration) was positively related to CYP1A2 activity as measured by the caffeine breath test (p < 0.01). Liver CYP1A2 activity was in turn negatively correlated with PCB-105:PCB-153 and PCB-118:PCB-153 congener ratios (p < 0.05). Hence, despite the relatively high body burden of PCBs and DLCs in this population, only smoking had a significant correlation with biomarkers of hepatic enzyme induction. Our data are consistent with smoking-induced liver CYP1A2 activity altering heme metabolism and increasing the biotransformation of mono-ortho PCB congeners.
Product Description
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®