Merck
  • The use of arginine brilliantgreen glucose peptone broth (ABGP medium) as a primary culture medium for Pseudomonas aeruginosa.
The use of arginine brilliantgreen glucose peptone broth (ABGP medium) as a primary culture medium for Pseudomonas aeruginosa.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. Serie B, Umwelthygiene, Krankenhaushygiene, Arbeitshygiene, praventive Medizin (1989-02-01)
R Schubert
ABSTRACT
A new medium is described for the isolation of P. aeruginosa organisms which have been damaged by chlorination. This medium utilises the fact that with arginine. P. aeruginosa produces a strongly alkaline reaction resulting in an easily identified colour change from gray-green to blue-violet. The indicators used for this purpose are bromothymol blue and cresol red. The medium may be used with either the membrane filter or the liquid enrichment technique. Incubation is at 37 degrees C for 24 or 48 h.
Sigma-Aldrich
Cresol red, JIS special grade
Sigma-Aldrich
Cresol Red sodium salt, indicator grade
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %