Skip to Content
Merck
All Photos(3)

Documents

114480

Sigma-Aldrich

Cresol Red sodium salt

indicator grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17NaO5S
CAS Number:
Molecular Weight:
404.41
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

indicator grade

form

solid

mp

250 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

λmax

425 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1cc(ccc1O)\C(=C2/C=CC(=O)C(C)=C2)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H18O5S.Na/c1-13-11-15(7-9-18(13)22)21(16-8-10-19(23)14(2)12-16)17-5-3-4-6-20(17)27(24,25)26;/h3-12,22H,1-2H3,(H,24,25,26);/q;+1/p-1/b21-16-;

InChI key

GBXGEGUVARLBPB-PLMZOXRSSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cresol Red has been used for checking the immobilized carbonic anhydrase-catalyzed reactions. It has also been used in the loading buffer for MHC (major histocompatibility complex) class I typing through PCR.

Biochem/physiol Actions

Cresol Red is used as a pH indicator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Horie et al.
Analytical biochemistry, 137(1), 80-87 (1984-02-01)
A highly sensitive quantitative method was developed to detect protons released or taken up upon ligand binding. A small change in pH due to proton release or uptake was detected by measuring the difference in the absorbance of a pH
[Fundamental studies on the measurement of urinary NAG and determination of the normal values for NAG activity in various age groups].
Y Iimura et al.
Rinsho byori. The Japanese journal of clinical pathology, 31(2), 179-184 (1983-02-01)
E Hodges et al.
Molecular pathology : MP, 50(3), 164-166 (1997-06-01)
Diagnostic immunoglobulin (Ig) polymerase chain reaction (PCR) clonality analyses need to be simple, reproducible, and rapid. Sucrose and cresol red (gel loading buffer reagents) were added to a routine IgH PCR reaction mix to obviate the need for adding gel
Paula C A Jerónimo et al.
The Analyst, 130(8), 1190-1197 (2005-07-16)
An optical biosensor based on immobilised carbonic anhydrase and its application to the determination of the anti-glaucoma agent acetazolamide by enzyme inhibition measurements, is described. The enzyme and a pH indicator dye, cresol red, were physically immobilised in overlapped sol-gel
K Elamrani et al.
Biochimica et biophysica acta, 727(1), 22-30 (1983-01-05)
The kinetics of H+/OH- diffusion across dimyristoyl phosphatidic acid bilayer membranes was measured by following the absorbance of the pH-sensitive indicator Cresol red (o-cresolsulfonphthalein) entrapped in single lamellar vesicles after rapidly changing the external pH in a stopped-flow apparatus. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service