Merck
  • [Inactivation of p53 leads to enhancement of tyrosine hydroxylase biosynthesis in the dopaminergic brain neurons].
[Inactivation of p53 leads to enhancement of tyrosine hydroxylase biosynthesis in the dopaminergic brain neurons].
Zhurnal evoliutsionnoi biokhimii i fiziologii (2013-06-26)
N A Dorofeeva, L S Nikitina, M V Glazova, O D Kirillova, E V Chernigovskaia
ABSTRACT
In the present work we analyzed a possibility of interaction of protein p53, family members of the ERK1/2 signaling cascade, and the transcription factor CREB in regulation of functional activity of dopaminergic neurons. There were considered neurons of Substantia nigra and Zona incerta of control rats and of rats injected intraperitoneally with chemical inhibitor of p53 Pifithrin-alpha blocking transcription activity ofproapoptotic protein p53. We have shown the p53 inactivation to lead to an increase in the content of tyrosine'hydroxylase both in cell bodies and in terminal parts of axons. At the same time, activity of the transcription factor CREB is enhanced in the brain dopaminergic neurons. No significant differences in the content of phospho-ERK1/2 kinases were revealed in the cell bodies at use of Pifithrin-alpha as compared with control group. Thus, we have shown that action of p53 on biosynthesis of tyrosine hydroxylase is of inhibitory character and seems to be mediated by the transcription factor CREB.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, JIS 1000, ≥99.8%
Sigma-Aldrich
Toluene, JIS 300, ≥99.8%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%