Merck
Stimuli-responsive peptide nanostructures at the fluid-fluid interface.
Methods in molecular biology (Clifton, N.J.) (2013-03-19)
Chun-Xia Zhao, Anton P J Middelberg
ABSTRACT
The self-organization of peptide-based nanostructures at a confined fluid-fluid interface, for example, the air-water or oil-water interface, is important in the context of stabilizing macroscopic soft-matter foams and emulsions. The unique ability to design interfacial nanostructures by controlling the subtle cooperativity that drives peptide self-assembly, and the ability to switch molecular cooperativity by facile triggers such as pH, opens new vistas for controlling macroscopic soft matter in industries as diverse as healthcare and industrial processing. Here we describe research aimed at developing new understanding into soft-matter formation and control, through variation of peptide sequence and bulk conditions. Macroscopic foaming and microfluidic emulsification studies prove particularly useful in visualizing and hence understanding the synergistic link between molecular design, mesoscopic interfacial properties, and bulk soft-matter stability.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, HPLC grade, 99.8%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%