Merck
  • Promoting immobilization and catalytic activity of horseradish peroxidase on mesoporous silica through template micelles.
Promoting immobilization and catalytic activity of horseradish peroxidase on mesoporous silica through template micelles.
Journal of colloid and interface science (2012-04-18)
Mi Mi Wan, Wei Gang Lin, Ling Gao, Hui Cheng Gu, Jian Hua Zhu
ABSTRACT
New concept on the promotion of immobilization and catalytic activity of enzyme on mesoporous silica through template micelles is proposed and realized in this paper. Proper P123 templates are controllable retained in the as-synthesized SBA-15, not only to anchor the horseradish peroxidase (HRP) guest, but also to establish the crowding-like microenvironment around the enzyme. The influence of retaining templates on the pore structure of SBA-15, immobilization, and catalytic activity of HRP is studied, and the possible process of template removal is proposed. Ethanol refluxing of 6 h is conformable to prepare the optimal mesoporous support characterized with the retained templates of about 8%. With the assistance of retained templates in SBA-15, up to 49 mg g(-1) of HRP can be immobilized, 100% more than that on calcined SBA-15. Furthermore, the thermal stability, the resistance of pH variation and denaturing agent urea, and the recycle usage of HRP immobilized are obviously elevated, paving a novel and low-cost route to develop enzyme catalysts.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 407, purified, non-ionic