Merck
Complete catalytic oxidation of o-xylene over CeO2 nanocubes.
Journal of environmental sciences (China) (2011-04-12)
Lian He, Yunbo Yu, Changbin Zhang, Hong He
ABSTRACT
The activities of CeO2 nanocubes calcined at different temperatures were tested for catalytic oxidation of o-xylene. Using CeO2 nanocubes as catalysts, complete catalytic oxidation of o-xylene was achieved below 210 degrees C. The CeO2 nanomaterials were characterized by means of BET, X-ray diffraction (XRD), transmission electron microscopy (TEM), and high-resolution transmission electron microscopy (HRTEM). From the TEM images, all CeO2 nanocubes displayed cubic morphology irrespective of calcination temperature. The HRTEM images revealed that these nanocubes were enclosed by reactive {001} planes, which may contribute to the intrinsically catalytic property of o-xylene oxidation. The higher activity of CeO2 nanocubes calcined at 550 degrees C than those calcined at above 550 degrees C was attributed to their smaller crystallite size and larger surface area. The influences of reaction conditions were also studied, which found that a higher reaction temperature was necessary for complete catalytic oxidation of o-xylene at higher weight hourly space velocity (WHSV) and o-xylene concentration.
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Xylenes, SAJ special grade, ≥80.0%
Sigma-Aldrich
Xylenes, SAJ first grade, ≥80.0%
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Xylenes, histological grade
Supelco
o-Xylene, suitable for HPLC, 98%