Merck
  • Production of Clostridium difficile toxin in a medium totally free of both animal and dairy proteins or digests.
Production of Clostridium difficile toxin in a medium totally free of both animal and dairy proteins or digests.
Proceedings of the National Academy of Sciences of the United States of America (2009-08-05)
Aiqi Fang, Donald F Gerson, Arnold L Demain
ABSTRACT
In the hope of developing a vaccine against Clostridium difficile based on its toxin(s), we have developed a fermentation medium for the bacterium that results in the formation of Toxin A and contains no meat or dairy products, thus obviating the problem of possible prion diseases. Particular preparations of hydrolyzed soy proteins, especially Soy Peptone A3, have been found to replace both the meat/dairy product tryptone in the preparation of working cell banks and seed media, and NZ-Soy BL4 does the same in the fermentation medium. These replacements yield even higher toxin titers.
Product Description
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Peptone from animal tissue, from meat
Millipore
Peptone Primatone® RL, suitable for microbiology
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Peptone from animal tissue, SAJ first grade
Millipore
Peptone from casein and other animal proteins, suitable for microbiology