Skip to Content
Merck
All Photos(1)

Key Documents

P0431

Millipore

Proteose Peptone

Enzymatic hydrolysate

Synonym(s):

Peptone from animal tissue

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

bovine
plant

Quality Level

form

powder

nitrogen analysis

10.0-15.5% total

loss

≤5% loss on drying

pH

6.6-7.6

application(s)

food and beverages
microbiology

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for use in antibiotic production and growth of fastidious microorganisms. Originally adapted for use in culture media for the production of bacterial toxins; a satisfactory nutrient for general culture needs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Priyadharshini Ramachandran et al.
Journal of microbiology and biotechnology, 23(3), 351-356 (2013-03-07)
A high beta-glucosidase (BGL)-producing strain, Stereum hirsutum, was identified and isolated and showed a maximum BGL activity (10.4 U/ml) when cultured with Avicel and tryptone as the carbon and nitrogen sources, respectively. In comparison with other BGLs, BGL obtained from
Takashi Kuda et al.
Food chemistry, 134(4), 1719-1723 (2013-02-28)
Listeria monocytogenes causes listeriosis in humans, mainly through the consumption of ready-to-eat foods such as cheese. Immunocompromised persons, the elderly, and pregnant women and their fetuses or newborns are at the highest risk for the infection. We examined the effects
Eliane Frimmersdorf et al.
Environmental microbiology, 12(6), 1734-1747 (2010-06-18)
In addition to transcriptome and proteome studies, metabolome analysis represents a third complementary approach to identify metabolic pathways and adaptation processes. In order to elucidate basic principles of metabolic versatility of Pseudomonas aeruginosa, we investigated the metabolome profiles of two
G Csaba et al.
Acta biologica Hungarica, 62(3), 228-234 (2011-08-16)
The aim of the experiments was to study the regulation of triiodothyronine (T3) production in the unicellular Tetrahymena. Untreated and troph-hormone treated specimen were prepared and in different timepoints T3 content was measured and compared by immunocytochemical flow cytometry. 0.1
Mesut Taskin et al.
International journal of food sciences and nutrition, 63(5), 597-602 (2011-12-06)
This study was performed to investigate the usability of chicken feather hydrolysate (Chicken feather peptone (CFP)) as substrate for mycelial biomass and extracellular polysaccharides (EPS) production from edible mushroom Morchella esculenta. The ability of CFP to support biomass and EPS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service