Merck
  • miR‑3613‑3p/MAP3K2/p38/caspase‑3 pathway regulates the heat‑stress‑induced apoptosis of endothelial cells.
miR‑3613‑3p/MAP3K2/p38/caspase‑3 pathway regulates the heat‑stress‑induced apoptosis of endothelial cells.
Molecular medicine reports (2021-07-20)
Jie Liu, Siya Xu, Shixin Liu, Bingguan Chen
ABSTRACT
Previous studies have identified microRNA (miRNA/miR)‑3613‑3p as a heat stress (HS)‑related miRNA in endothelial cells that can lead to apoptosis. However, the mechanism underlying the miR‑3613‑3p‑mediated apoptosis of HS‑exposed endothelial cells remains unclear. In the present study, western blot analysis and reverse transcription‑quantitative PCR were used to determine protein and miRNA expression levels, respectively. Annexin V‑fluorescein isothiocyanate/propidium iodide staining, caspase‑3 activity measurements and DNA fragmentation assays were performed to detect apoptosis. To evaluate whether mitogen‑activated protein kinase kinase kinase 2 (MAP3K2) was a direct target of miR‑3613‑3p, a luciferase reporter assay was performed. In addition, transient transfection was used to carry out loss‑ and gain‑of‑function experiments. The results revealed that miR‑3613‑3p expression was reduced in human umbilical vein endothelial cells (HUVECs) following HS, which led to apoptosis. Mechanistically, following HS, a decrease in miR‑3613‑3p binding to the 3'‑untranslated region of MAP3K2 directly upregulated its expression, and the downstream p38 and caspase‑3 pathways, thereby leading to apoptosis. Taken together, the results of the present study demonstrated that HS suppressed miR‑3613‑3p expression, which activated the MAP3K2/p38/caspase‑3 pathway, leading to the apoptosis of HUVECs. In conclusion, the miR‑3613‑3p/MAP3K2/p38/caspase‑3 pathway may serve an indispensable role in regulating the progression of apoptosis, indicating a regulatory role of miR‑3613‑3p in the pathophysiology of HS‑exposed endothelial cells.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Fc, HRP conjugate, 0.8 mg/mL, Chemicon®
Sigma-Aldrich
Anti-p38 MAP Kinase, Activated (Diphosphorylated p38) antibody, Mouse monoclonal, clone P38-TY, purified from hybridoma cell culture, buffered aqueous solution
Sigma-Aldrich
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated/monophosphorylated (Phosphothreonine ERK-1&2) antibody produced in mouse, ~2 mg/mL, clone ERK-PT115, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-p38 (Thr180/Tyr182) Antibody, clone 6E5.2, clone 6E5.2, from mouse
Sigma-Aldrich
Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-phospho JNK (Thr183, Tyr185) Antibody, clone 3F7, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells
Sigma-Aldrich
Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid