Skip to Content
Merck
All Photos(4)

Documents

M8159

Sigma-Aldrich

Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse

clone MAPK-YT, ascites fluid

Synonym(s):

Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160410
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MAPK-YT, monoclonal

mol wt

antigen ERK-1 44 kDa
antigen ERK-2 42 kDa

contains

15 mM sodium azide

species reactivity

human, Caenorhabditis elegans, Xenopus, Drosophila, hamster, rat, bovine, mouse, yeast

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:10,000 using rat brain extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

MAP kinase (MAPK, mitogen-activated protein kinase is also termed as ERK, extracellular regulated protein kinase). Molecular cloning has established that MAP kinase (ERKs) consists of at least four isoforms: ERK-1 (p44mapk), ERK-2 (p42mapk), ERK-3, and ERK-5. MAP kinase isoforms appear to be widely expressed in the central nervous system, thymus, spleen, heart, lung and kidney. It is also expressed at high levels in PC12 cells and in fibroblasts. MAPK1 is located on human chromosome 22q11. MAPK3 is mapped to human chromosome 16p11.

Specificity

The antibody reacts specifically with the diphosphorylated form of MAP kinase (ERK-1 and ERK-2). It does not recognize the non-phosphorylated or the monophosphorylated forms of MAP kinase or the diphosphorylated forms of JNK and p38 MAP kinase. The epitope recognized by the antibody contains the phosphorylated threonine and tyrosine residues within the regulatory site of active MAP kinase.

Immunogen

synthetic peptide HTGFLpTEpYVAT corresponding to the phosphorylated form of the ERK-activation loop.

Application

Antibodies that react specifically with the active form of MAP kinase are useful for the study of the specific activation requirements, differential tissue expression, and intracellular localization of the active form of MAP kinase in normal and neoplastic tissue.
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) may be used for the localization of the active, dually-phosphorylated, form of MAP kinase using various immunochemical assays such as immunoblotting of cultured cells and tissue extracts, ELISA, immunocytochemistry, immunoprecipitation, and in immunohistochemistry (formalin and formaldehyde-fixed sections). Reactivity has been observed with human, bovine, rat, mouse, Drosophila, Spodoptera frugiperda, and yeast.

Monoclonal Anti-MAP Kinase, activated (Diphosphorylated ERK-1&2) antibody has been used in plasmid and transient transfection and western blotting.

Biochem/physiol Actions

MAP kinase (MAPK, mitogen-activated protein kinase) plays a crucial role in various signal transduction pathways leading signals of growth factor, as well as G protein-coupled receptors to their intracellular targets. MAP kinase was shown to regulate several cellular processes among them proliferation, differentiation, cellular morphology and oncogenesis. Activation of ERK-1 and ERK-2 in mitogen-stimulated cells is directly mediated by MAP kinase kinase (MAPKK or MEK), a dual-specificity protein kinase, which phosphorylates both threonine and tyrosine residues in the regulatory sites of MAP kinase. Following activation, MAP kinase phosphorylates several nuclear targets, including transcription factors as well as membrane and cytoskeletal proteins. Termination of MAP kinase signalling appears to be mediated by MAP kinase phosphatase, MKP-1, a dual specificity Thr/Tyr phosphatase which dephosphorylates and inactivates MAP kinase.

Physical form

The product is provided as ascites fluid containing 15 mM sodium azide as a preservative.

Storage and Stability

Store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression and genotyping studies implicate the interleukin 7 receptor in the pathogenesis of primary progressive multiple sclerosis.
Booth D R, et al.
Journal of Molecular Medicine, 83(10), 822-830 (2005)
The phosphatidylinositol-3 kinase/Akt pathway mediates VEGF?s neuroprotective activity and induces blood brain barrier permeability after focal cerebral ischemia
Kilic E, et al.
Faseb Journal, 20(8), 1185-1187 (2006)
Mitogen-Activated Protein (MAP) Kinase Pathways: Regulation and Physiological Functions.
Gray P, et al.
Endocrine Reviews, 22(2), 153-183 (2001)
Mary Truscott et al.
PLoS genetics, 10(7), e1004493-e1004493 (2014-07-25)
The importance of microRNAs in the regulation of various aspects of biology and disease is well recognized. However, what remains largely unappreciated is that a significant number of miRNAs are embedded within and are often co-expressed with protein-coding host genes.
Mitogen activated protein (MAP) kinase signal transduction pathways and novel anti-inflammatory targets.
Hommes D W, et al.
Gut, 52(1), 144-151 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service