Merck
  • Post-translational polymodification of β1-tubulin regulates motor protein localisation in platelet production and function.
Post-translational polymodification of β1-tubulin regulates motor protein localisation in platelet production and function.
Haematologica (2020-12-18)
Abdullah O Khan, Alexandre Slater, Annabel Maclachlan, Phillip L R Nicolson, Jeremy A Pike, Jasmeet S Reyat, Jack Yule, Rachel Stapley, Julie Rayes, Steven G Thomas, Neil V Morgan
ABSTRACT
In specialised cells, the expression of specific tubulin isoforms and their subsequent post-translational modifications drive and coordinate unique morphologies and behaviours. The mechanisms by which β1-tubulin, the platelet and megakaryocyte (MK) lineage restricted tubulin isoform, drives platelet production and function remains poorly understood. We investigated the roles of two key post-translational tubulin polymodifications (polyglutamylation and polyglycylation) on these processes using a cohort of thrombocytopenic patients, human induced pluripotent stem cell (iPSC) derived MKs, and healthy human donor platelets. We find distinct patterns of polymodification in MKs and platelets, mediated by the antagonistic activities of the cell specific expression of Tubulin Tyrosine Ligase Like (TTLLs) and Cytosolic Carboxypeptidase (CCP) enzymes. The resulting microtubule patterning spatially regulates motor proteins to drive proplatelet formation in megakaryocytes, and the cytoskeletal reorganisation required for thrombus formation. This work is the first to show a reversible system of polymodification by which different cell specific functions are achieved.
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-monoglycylated Tubulin Antibody, clone TAP 952, clone TAP 952, from mouse
Sigma-Aldrich
Anti-pan polyglycylated Tubulin Antibody, clone AXO 49, clone AXO 49, from mouse
Sigma-Aldrich
Anti-Tubulin, Polyglutamylated antibody, Mouse monoclonal, clone B3, purified from hybridoma cell culture