Skip to Content
Merck
All Photos(1)

Key Documents

MABS276

Sigma-Aldrich

Anti-pan polyglycylated Tubulin Antibody, clone AXO 49

clone AXO 49, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AXO 49, monoclonal

species reactivity

Apis, porcine, snail, pig, sea urchin, protista, sheep, mouse, lemur, Drosophila, Paramecium, trout

should not react with

Euglena, human (cilia)

technique(s)

dot blot: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Several post-translational modifications of tubulin have been identified in eukaryotic cells. Glycylation is a poly-modification which occurs as lateral branching of glycine chains of variable length identified in axonemal tubulin of Paramecium cilia. This modification has been detected on tubulin and/or cilia/flagella of many unicellular and pluricellular organisms. The differential distribution of distinct polyglycylated tubulin isoforms between cytoplasmic and axonemal compartments indicates that polyglycylation levels of tubulin is highly regulated at the cell level. The AXO49 antibody is reactive mainly on axonemal tubulin of motile cilia and flagella which are polyglycylated. In some cases, reactivity may also be observed on other very stable microtubule networks. This antibody is useful for cilia and flagella detection.

Specificity

This antibody recognizes the lateral connecting branches of a polymer, which contains at least 3 glycine residues. This antibody demonstrates cross reactivity against polyglycylated tubulin, as well as with other polyglycylated proteins .

Immunogen

Insoluble fraction of Paramecium cilia (Paramecium axonemes)

Application

Detect Tubulin using this mouse monoclonal antibody, Anti-pan polyglycylated Tubulin Antibody, clone AXO 49 validated for use in western blotting, Immunofluorescence & Dot Blot.

Quality

Evaluated by Western Blotting on total cytoskeletal proteins of Paramecium. : A 1:50,000 dilution of this antibody detected polyglycylated Tubulin in 10 µg of Paramecium total cytoskeletal proteins.

Target description

~55 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzanne H Hodge et al.
Biology open, 10(8) (2021-08-07)
Primary cilia are compartmentalised from the rest of the cell by a ciliary gate comprising transition fibres and a transition zone. The ciliary gate allows the selective import and export of molecules such as transmembrane receptors and transport proteins. These
Harrison D Pravder et al.
International journal of molecular sciences, 23(6) (2022-03-26)
A useful model for determining the mechanisms by which actin and actin binding proteins control cellular architecture is the Drosophila melanogaster process of spermatogenesis. During the final step of spermatogenesis, 64 syncytial spermatids individualized as stable actin cones move synchronously
Jennifer Lennon et al.
Frontiers in genetics, 13, 943197-943197 (2022-07-26)
Axonemal dynein motors are large multi-subunit complexes that drive ciliary movement. Cytoplasmic assembly of these motor complexes involves several co-chaperones, some of which are related to the R2TP co-chaperone complex. Mutations of these genes in humans cause the motile ciliopathy
Haibo Xie et al.
Journal of molecular cell biology, 14(7) (2022-08-19)
Meiosis is essential for evolution and genetic diversity in almost all sexual eukaryotic organisms. The mechanisms of meiotic recombination, such as synapsis, have been extensively investigated. However, it is still unclear whether signals from the cytoplasm or even from outside
Sara Molina-Gil et al.
Nature communications, 14(1), 5730-5730 (2023-09-16)
The re-use of genes in new organs forms the base of many evolutionary novelties. A well-characterised case is the recruitment of the posterior spiracle gene network to the Drosophila male genitalia. Here we find that this network has also been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service