Merck
  • Diet-dependent regulation of TGFβ impairs reparative innate immune responses after demyelination.
Diet-dependent regulation of TGFβ impairs reparative innate immune responses after demyelination.
Nature metabolism (2021-02-24)
Mar Bosch-Queralt, Ludovico Cantuti-Castelvetri, Alkmini Damkou, Martina Schifferer, Kai Schlepckow, Ioannis Alexopoulos, Dieter Lütjohann, Christian Klose, Lenka Vaculčiaková, Takahiro Masuda, Marco Prinz, Kathryn M Monroe, Gilbert Di Paolo, Joseph W Lewcock, Christian Haass, Mikael Simons
ABSTRACT
Proregenerative responses are required for the restoration of nervous-system functionality in demyelinating diseases such as multiple sclerosis (MS). Yet, the limiting factors responsible for poor CNS repair are only partially understood. Here, we test the impact of a Western diet (WD) on phagocyte function in a mouse model of demyelinating injury that requires microglial innate immune function for a regenerative response to occur. We find that WD feeding triggers an ageing-related, dysfunctional metabolic response that is associated with impaired myelin-debris clearance in microglia, thereby impairing lesion recovery after demyelination. Mechanistically, we detect enhanced transforming growth factor beta (TGFβ) signalling, which suppresses the activation of the liver X receptor (LXR)-regulated genes involved in cholesterol efflux, thereby inhibiting phagocytic clearance of myelin and cholesterol. Blocking TGFβ or promoting triggering receptor expressed on myeloid cells 2 (TREM2) activity restores microglia responsiveness and myelin-debris clearance after demyelinating injury. Thus, we have identified a druggable microglial immune checkpoint mechanism regulating the microglial response to injury that promotes remyelination.
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Fenofibrate, ≥99%, powder
Sigma-Aldrich
Lipid Mixture 1, Chemically Defined, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Liquid nitrogen cooled mini mortar, pkg of 1 ea
Sigma-Aldrich
L-α-Lysophosphatidylcholine from egg yolk, ≥99%, Type I, powder
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Monoclonal Anti-ALDH1L1 antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution