Merck
  • A chemical screening approach reveals that indole fluorescence is quenched by pre-fibrillar but not fibrillar amyloid-beta.
A chemical screening approach reveals that indole fluorescence is quenched by pre-fibrillar but not fibrillar amyloid-beta.
Bioorganic & medicinal chemistry letters (2009-07-31)
Ashley A Reinke, Han Yiau Seh, Jason E Gestwicki
ABSTRACT
Aggregated amyloid-beta (Abeta) peptide is implicated in the pathology of Alzheimer's disease. In vitro and in vivo, these aggregates are found in a variety of morphologies, including globular oligomers and linear fibrils, which possess distinct biological activities. However, known chemical probes, including the dyes thioflavin T and Congo Red, appear to lack selectivity for specific amyloid structures. To identify molecules that might differentiate between these architectures, we employed a fluorescence-based interaction assay to screen a collection of 68 known Abeta ligands against pre-formed oligomers and fibrils. In these studies, we found that the fluorescence of five indole-based compounds was selectively quenched ( approximately 15%) in the presence of oligomers, but remained unchanged after addition of fibrils. These results suggest that indoles might be complementary to existing chemical probes for studying amyloid formation in vitro.
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Sigma-Aldrich
Rosmarinic acid, 96%
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
3-Hydroxybenzaldehyde, ≥99%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Indole-3-carboxaldehyde, 97%
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%
Sigma-Aldrich
Anthrone, ACS reagent, 97%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)