Merck
  • Structure-activity correlations for beta-phenethylamines at human trace amine receptor 1.
Structure-activity correlations for beta-phenethylamines at human trace amine receptor 1.
Bioorganic & medicinal chemistry (2008-07-08)
Anita H Lewin, Hernán A Navarro, S Wayne Mascarella
ABSTRACT
A cell line in which RD-HGA16 cells were stably transfected with the hTAAR 1 receptor was created and utilized to carry out a systematic evaluation of a series of beta-phenethylamines. Fair agreement was observed with data obtained for aryl and ethylene chain substituted analogs in an AV12-664 cell line in which hemagglutinin-tagged hTAAR 1 was stably co-expressed with rat G alpha(s). Analogs with multiple substituents as well as analogs with bulky groups were found to be partial agonists. Analogs in which the primary amino group was converted to a secondary or a tertiary amino group by N-methylation were also partial agonists. Comparative Molecular Field Analysis (CoMFA) using the potency data yielded a regression coefficient r(2) of 0.824. The steric field contribution to the model was 61% with the balance (39%) contributed by the electrostatic field. The collective results suggest that increasing steric bulk both at the amino nitrogen, particularly by N-dimethylation, and at the 4-position of the aromatic ring leads to low efficacy ligands.
Product Description
Supelco
2-Phenethylamine, analytical standard
Sigma-Aldrich
N-Methyl-phenethylamine, 99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
(R)-(+)-β-Methylphenethylamine, 99%
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
2-(p-Tolyl)ethylamine, 97%
Sigma-Aldrich
N,N-Dimethylphenethylamine, 98%
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
3-Methoxyphenethylamine, 97%