Skip to Content
Merck
All Photos(1)

Key Documents

523801

Sigma-Aldrich

N,N-Dimethylphenethylamine

98%

Synonym(s):

2-Phenylethyl-N,N-dimethylamine, Dimethyl(2-phenylethyl)amine, N,N-Dimethyl-β-phenethylamine, N,N-Dimethyl-2-phenylethylamine, N,N-Dimethylbenzeneethanamine, N,N-Dimethylphenylethylamine, N-(2-Phenylethyl)dimethylamine, N-Phenethyldimethylamine, [(Dimethylamino)ethyl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2N(CH3)2
CAS Number:
Molecular Weight:
149.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.502 (lit.)

bp

207-212 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

CN(C)CCc1ccccc1

InChI

1S/C10H15N/c1-11(2)9-8-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3

InChI key

TXOFSCODFRHERQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Inoue et al.
Journal of neurochemistry, 44(1), 210-216 (1985-01-01)
N-[methyl-14C]N,N-dimethylphenylethylamine (DMPEA) was synthesized and its availability as a selective radiotracer for in vivo measurement of mouse brain monoamine oxidase (MAO) activity was examined. Relatively high incorporation of labelled DMPEA into brain (about 10% of the injected dose/per gram of
S Shirolkar et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 11(1), 41-47 (1995-01-01)
In an effort to develop a long-term topically active tear stimulant, it was important to determine if tolerance developed following the repeated instillation of N,N-Dimethyl-2-phenylethylamine hydrochloride (AF2975) to the albino rabbit eye. New Zealand white rabbits were administered AF2975 (0.15%)
R D Schoenwald et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 14(3), 253-262 (1998-07-22)
Acini cells were obtained from the lacrimal gland of the white New Zealand rabbit. Following isolation and purification, the cells were used to study the uptake of N,N'-dimethyl-2-phenylethylamine HCl (AF2975), which was found to be sodium- and proton-independent, but energy-dependent.
H Shinotoh et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 28(6), 1006-1011 (1987-06-01)
Carbon-11-labeled N,N-dimethylphenylethylamine ([11C]DMPEA) was synthesized by the reaction of N-methylphenylethylamine with [11C]methyl iodide. This newly synthesized radiotracer was developed for the purpose of in vivo measurement of monoamine oxidase-B activity in the brain using a metabolic trapping method. Initially, biodistribution
R D Schoenwald et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 13(2), 151-161 (1997-04-01)
Fourteen dry eye volunteers placed one to two drops of 0.15% AF2975 (N,N-dimethyl-2-phenylethylamine HCl) in one eye and the vehicle in their other eye four times a day for 21 days. AF2975 is a sigma agonist known to stimulate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service