Merck
  • Synthesis and anti-Trypanosoma cruzi activity of derivatives from nor-lapachones and lapachones.
Synthesis and anti-Trypanosoma cruzi activity of derivatives from nor-lapachones and lapachones.
Bioorganic & medicinal chemistry (2008-04-02)
Eufrânio N da Silva Júnior, Maria Cecília B V de Souza, Michelle C Fernandes, Rubem F S Menna-Barreto, Maria do Carmo F R Pinto, Francisco de Assis Lopes, Carlos Alberto de Simone, Carlos Kleber Z Andrade, Antônio V Pinto, Vitor F Ferreira, Solange L de Castro
ABSTRACT
New naphthoquinone derivatives were synthesized and assayed against bloodstream trypomastigote forms of Trypanosoma cruzi, the etiological agent of Chagas' disease. The compounds were rationalized based on hybrid drugs and appear as important compounds against this parasite. From nor-lapachol were prepared five substituted ortho-naphthofuranquinones, a non-substituted para-naphthofuranquinone, a new oxyrane and an azide and from alpha-lapachone a new non-substituted para-naphthofuranquinone. Other five substituted ortho-naphthofuranquinones recently designed as cytotoxic, were also evaluated. The most active compounds were the ortho naphthofuranquinones 3-(4-methoxyphenylamino)-2,3-dihydro-2,2-dimethylnaphtho[1,2-b]furan-4,5-dione and 3-(3-nitrophenylamino)-2,3-dihydro-2,2-dimethylnaphtho[1,2-b]furan-4,5-dione with trypanocidal activity higher than that of benznidazole, the standard drug. The compounds were rationalized based on hybrid drugs and appear as important compounds against T. cruzi. The trypanocidal activity of these substances endowed with redox properties representing a good starting point for a medicinal chemistry program aiming the chemotherapy of Chagas' disease.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet Solution