Merck
  • 8p11.23 Amplification in Breast Cancer: Molecular Characteristics, Prognosis and Targeted Therapy.
8p11.23 Amplification in Breast Cancer: Molecular Characteristics, Prognosis and Targeted Therapy.
Journal of clinical medicine (2020-09-30)
Ioannis A Voutsadakis
ABSTRACT
Amplification of the locus 8p11.23 has been observed in cancer and genes of this locus, including ZNF703 (Zinc finger protein 703), NSD3 (Nuclear receptor binding SET domain protein 3) and FGFR1 (Fibroblast growth factor receptor 1), have been put forward as dominant oncogenes conferring pathophysiologic benefit in cancers with amplifications. However, there is no consensus on the importance of each of them or any other genes of the amplicon or even a consensus on which genes are part of the amplicon. Publicly available data were used to characterize the locus amplified at 8p11.23 and derive information on each of the genes and roles as oncogenes. The frequency of the amplifications in the locus was examined in the cBioportal platform, and expression levels of the amplicon genes in amplified cases were derived from genomic studies reported in the platform. Examination of the influence of mRNA expressions of each gene of the locus for Recurrence-free survival in breast cancer was performed using K-M plotter. The 8p11.23 amplicon is present in higher frequency in squamous cell lung carcinomas, breast cancers and bladder carcinomas and is only rarely observed in other cancers. The most frequently amplified genes within the amplicon vary between different types of cancers. In breast cancer, amplified cases are most commonly of the luminal B type. Amplified genes are not always over-expressed and there is a low correlation of amplification with over-expression in amplicon genes with variation between genes. The presence of the amplicon does not influence the aneuploidy score or the tumor mutation burden of breast cancers. Regarding prognosis, the two genes of the amplicon whose mRNA hyper-expression portends adverse relapse-free survival in breast cancer are EIF4EBP1 (Eukaryotic transcription initiation factor 4E binding protein 1) and LSM1 (LSM1 homolog, mRNA degradation associated). Besides the previously proposed genes to play a role as dominant oncogenes in the 8p11.23 cancer amplified locus, other genes may also be important in breast cancer based on the high correlation of their amplification and mRNA expression and adverse prognosis conferred by over-expression, consistent with an oncogenic role.
Sigma-Aldrich
Anti-DDHD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-ASH2L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NSD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ZNF703 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-LETM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NSD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-BAG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DDHD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GOT1L1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ADRB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-RAB11FIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-TACC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RAB11FIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-ERLIN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-BRF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-STAR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
Anti-STAR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-PROSC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-PROSC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-RAB11FIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
Sigma-Aldrich
Anti-ADGRA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FGFR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution