Skip to Content
Merck
All Photos(3)

Key Documents

HPA023646

Sigma-Aldrich

Anti-PROSC antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Proline synthetase co-transcribed bacterial homolog protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VPNLFMLETVDSVKLADKVNSSWQRKGSPERLKVMVQINTSGEESKHGLPPSETIAIVEHINAKCPNLEFVGLMT

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PROSC(11212)

General description

PROSC (proline synthetase co-transcribed) is a gene which codes for a soluble cytoplasmic protein. This gene is located on human chromosome 8p11.2.. Its cDNA is 2530 bp long, with eight exons that includes 275 amino acids.

Immunogen

Proline synthetase co-transcribed bacterial homolog protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PROSC antibody has been used in immunoblot analysis.

Biochem/physiol Actions

PROSC (proline synthetase co-transcribed) participates in intracellular homeostatic regulation of PLP (pyridoxal 5′-phosphate). It plays an important role in cellular function and bacterial PROSC may participate in proline synthesis. In cytoplasm, PROSC may stimulate the sensitivity to ara-C (cytosine arabinoside). Mutations in PROSC can distort cellular pyridoxal phosphate homeostasis and lead to vitamin-B6-dependent epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76262

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and characterization of human and mouse PROSC (proline synthetase co-transcribed) genes.
Ikegawa S, et al.
Journal of Human Genetics, 44(5), 337-342 (1999)
Gene expression of PROSC (proline synthetase co-transcribed) in related to cytosine arabinoside sensitivity in human leukemia.
Fujimaki SI, et al.
Journal of electrophoresis, 52(3), 53-56 (2008)
Mutations in PROSC disrupt cellular pyridoxal phosphate homeostasis and cause vitamin-B6-dependent epilepsy.
Darin N, et al.
American Journal of Human Genetics, 99(6), 1325-1337 (2016)
Niklas Darin et al.
American journal of human genetics, 99(6), 1325-1337 (2016-12-03)
Pyridoxal 5'-phosphate (PLP), the active form of vitamin B6, functions as a cofactor in humans for more than 140 enzymes, many of which are involved in neurotransmitter synthesis and degradation. A deficiency of PLP can present, therefore, as seizures and
Maitou Pal et al.
JIMD reports, 59(1), 32-41 (2021-05-13)
Pyridoxine-dependent epilepsy (PDE) is a relatively rare subgroup of epileptic disorders. They generally present in infancy as an early onset epileptic encephalopathy or seizures, refractory to standard treatments, with rapid and variable responses to vitamin B6 treatment. Whole exome sequencing of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service