Merck
  • Qualitative Factor-Based Comparison of NMR, Targeted and Untargeted GC-MS and LC-MS on the Metabolomic Profiles of Rioja and Priorat Red Wines.
Qualitative Factor-Based Comparison of NMR, Targeted and Untargeted GC-MS and LC-MS on the Metabolomic Profiles of Rioja and Priorat Red Wines.
Foods (Basel, Switzerland) (2020-10-03)
Dimitrios Kioroglou, Albert Mas, Maria C Portillo
ABSTRACT
Wine origin and ageing are two factors related to wine quality which in turn is associated to wine metabolome. Currently, new metabolomic techniques and proper statistics procedures allow accurate profiling of wine metabolome. Thus, the main goal was to evaluate different metabolomic methodologies on their ability to provide patterns on the wine metabolome based on selected factors, such as ageing of barrel-aged wine (factor time), prior usage of the barrels (factor barrel-type), and differences between wine ageing in barrels or glass bottles (factor bottled-wine). In the current study, we implement NMR, targeted and untargeted GC-MS and LC-MS metabolomic analytical techniques so as to gain insights into the volatile and nonvolatile wine metabolome composition of red wines from two cellars located in the only two Spanish Qualified Appellations of Origin; DOQ Priorat and DOCa Rioja regions. Overall, 95 differentially significant metabolites were identified facilitating the evaluation of the analytical methodologies performance and finding common trends of those metabolites depending on the considered factor. The results did not favor NMR as an effective technique on the current dataset whereas suggested LC-MS as an adequate technique for revealing differences based on the factor time, targeted GC-MS on the factor barrel-type, and untargeted GC-MS on the factor bottled-wine. Thus, a combination of different metabolomic techniques is necessary for a complete overview of the metabolome changes. These results ease the selection of the correct methodology depending on the specific factor investigated.
Sigma-Aldrich
2-(4-Biphenylyl)-6-phenylbenzoxazole, BioReagent, suitable for scintillation, ≥98.0% (TLC)
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Supelco
4-Ethylguaiacol, analytical standard
Sigma-Aldrich
4-Ethylphenol, 99%