Merck
  • Absolute Quantification of the Central Carbon Metabolome in Eight Commonly Applied Prokaryotic and Eukaryotic Model Systems.
Absolute Quantification of the Central Carbon Metabolome in Eight Commonly Applied Prokaryotic and Eukaryotic Model Systems.
Metabolites (2020-02-26)
Lisa M Røst, Lilja Brekke Thorfinnsdottir, Kanhaiya Kumar, Katsuya Fuchino, Ida Eide Langørgen, Zdenka Bartosova, Kåre Andre Kristiansen, Per Bruheim
ABSTRACT
Absolute quantification of intracellular metabolite pools is a prerequisite for modeling and in-depth biological interpretation of metabolomics data. It is the final step of an elaborate metabolomics workflow, with challenges associated with all steps-from sampling to quantifying the physicochemically diverse metabolite pool. Chromatographic separation combined with mass spectrometric (MS) detection is the superior platform for high coverage, selective, and sensitive detection of metabolites. Herein, we apply our quantitative MS-metabolomics workflow to measure and present the central carbon metabolome of a panel of commonly applied biological model systems. The workflow includes three chromatographic methods combined with isotope dilution tandem mass spectrometry to allow for absolute quantification of 68 metabolites of glycolysis, the pentose phosphate pathway, the tricarboxylic acid cycle, and the amino acid and (deoxy) nucleoside pools. The biological model systems; Bacillus subtilis, Saccharomyces cerevisiae, two microalgal species, and four human cell lines were all cultured in commonly applied culture media and sampled in exponential growth phase. Both literature and databases are scarce with comprehensive metabolite datasets, and existing entries range over several orders of magnitude. The workflow and metabolite panel presented herein can be employed to expand the list of reference metabolomes, as encouraged by the metabolomics community, in a continued effort to develop and refine high-quality quantitative metabolomics workflows.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Millipore
Tryptone, Pancreatic digest of casein, Suitable for microbiology
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Millipore
YPD Broth, suitable for microbiology, NutriSelect® Basic
Millipore
Yeast Extract, suitable for microbiology
Millipore
PVDF Membrane Filter, 5.0 μm Pore Size, Durapore®, filter diam. 47 mm, hydrophilic, white
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture