Merck
  • Transient Receptor Potential V Channels Are Essential for Glucose Sensing by Aldolase and AMPK.
Transient Receptor Potential V Channels Are Essential for Glucose Sensing by Aldolase and AMPK.
Cell metabolism (2019-06-18)
Mengqi Li, Chen-Song Zhang, Yue Zong, Jin-Wei Feng, Teng Ma, Meiqin Hu, Zhizhong Lin, Xiaotong Li, Changchuan Xie, Yaying Wu, Dong Jiang, Ying Li, Cixiong Zhang, Xiao Tian, Wen Wang, Yanyan Yang, Jie Chen, Jiwen Cui, Yu-Qing Wu, Xin Chen, Qing-Feng Liu, Jianfeng Wu, Shu-Yong Lin, Zhiyun Ye, Ying Liu, Hai-Long Piao, Li Yu, Zhuan Zhou, Xiao-Song Xie, D Grahame Hardie, Sheng-Cai Lin
ABSTRACT
Fructose-1,6-bisphosphate (FBP) aldolase links sensing of declining glucose availability to AMPK activation via the lysosomal pathway. However, how aldolase transmits lack of occupancy by FBP to AMPK activation remains unclear. Here, we show that FBP-unoccupied aldolase interacts with and inhibits endoplasmic reticulum (ER)-localized transient receptor potential channel subfamily V, inhibiting calcium release in low glucose. The decrease of calcium at contact sites between ER and lysosome renders the inhibited TRPV accessible to bind the lysosomal v-ATPase that then recruits AXIN:LKB1 to activate AMPK independently of AMP. Genetic depletion of TRPVs blocks glucose starvation-induced AMPK activation in cells and liver of mice, and in nematodes, indicative of physical requirement of TRPVs. Pharmacological inhibition of TRPVs activates AMPK and elevates NAD+ levels in aged muscles, rejuvenating the animals' running capacity. Our study elucidates that TRPVs relay the FBP-free status of aldolase to the reconfiguration of v-ATPase, leading to AMPK activation in low glucose.
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 10,000
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Diazoxide
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Dounce tissue grinder pestle, Large clearance, working volume 7 mL
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
RN-1747, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Phenformin hydrochloride, analytical standard
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
L-Lactate Dehydrogenase (LDHA), from human, recombinant, expressed in E. coli, aqueous solution
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
Carvacrol, ≥98%, FCC, FG
Sigma-Aldrich
GSK1016790A, ≥98% (HPLC)
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Concanamycin A, ≥70% (HPLC)