Merck
  • Derivation and propagation of embryonic stem cells in serum- and feeder-free culture.
Derivation and propagation of embryonic stem cells in serum- and feeder-free culture.
Methods in molecular biology (Clifton, N.J.) (2006-07-19)
Jennifer Nichols, Qi-Long Ying
ABSTRACT
The availability of murine embryonic stem (ES) cells has revolutionized the study of mammalian development and disease. We recently developed a culture medium that has enabled us to identify the essential signaling pathways required for maintenance of pluripotency in vitro. Addition of leukemia inhibitory factor and bone morphogenetic protein4 to this medium is sufficient to activate the signal transducer and activator of transcription3 and mammalian homolog of Drosophila mothers against decapentaplegic pathways, respectively. We have successfully derived and propagated ES cells in the absence of feeder cells and serum. This chapter describes a simple protocol for efficient derivation and maintenance of ES cells from embryos of the 129 and C57B1/6 strains of mice.
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Chicken Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
NDiff Neuro-27 Medium Supplement (100x), NDiff Neuro-27 Medium Supplement is a serum-free supplement containing antioxidants & other factors that has been developed for the in vitro propagation & maintenance of undifferentiated murine embryonic stem (ES) cells in serum free medium.
Sigma-Aldrich
NDiff Neuro-2 Medium Supplement (200x), NDiff Neuro-2 Medium Supplement is a serum-free, N2-like supplement for the in vitro differentiation of murine embryonic stem (ES) cells into post-mitotic neurons particularly via monolayer differentiation.
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%