Skip to Content
Merck
All Photos(1)

Key Documents

SCM013

Sigma-Aldrich

NDiff Neuro-27 Medium Supplement (100x)

NDiff Neuro-27 Medium Supplement is a serum-free supplement containing antioxidants & other factors that has been developed for the in vitro propagation & maintenance of undifferentiated murine embryonic stem (ES) cells in serum free medium.

Synonym(s):

NDiff Medium Supplement, Neuro-27 Medium Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Chemicon®
Millipore

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)

shipped in

dry ice

General description

NDiff Neuro-27 Medium Supplement is a serum-free supplement containing antioxidants and other factors that has been specifically developed for the in vitro propagation and maintenance of undifferentiated murine embryonic stem (ES) cells in serum free medium (Nichols J, 2006). This product can also be used to differentiate murine ES cells into post-mitotic neurons particularly via monolayer differentiation (Ying QL, 2003).

Physical form

NDiff Neuro-27 Medium Supplement is supplied as an aqueous solution. Optimal working concentration of the medium supplement must be determined for specific uses and cell types. In most instances, a 1:40-1:200 dilution is appropriate. Typically, Neuro-27 is diluted 1:100 for self renewal and neural differentiation of mouse ES cells.

Storage and Stability

Maintain at -20 °C until expiration date. Protect from light. Once thawed, aliquot unused portion and store at -20 °C until future use. Once added to the cell culture medium, the product is stable at 4 °C for four weeks.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Defined conditions for neural commitment and differentiation
Ying, Qi-Long and Smith, Austin G
Test, 365, 327-341 (2003)
Qi-Long Ying et al.
Nature biotechnology, 21(2), 183-186 (2003-01-14)
Mouse embryonic stem (ES) cells are competent for production of all fetal and adult cell types. However, the utility of ES cells as a developmental model or as a source of defined cell populations for pharmaceutical screening or transplantation is
Jennifer Nichols et al.
Methods in molecular biology (Clifton, N.J.), 329, 91-98 (2006-07-19)
The availability of murine embryonic stem (ES) cells has revolutionized the study of mammalian development and disease. We recently developed a culture medium that has enabled us to identify the essential signaling pathways required for maintenance of pluripotency in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service