Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead(II) sulfate
Synonym(s):
Anglesite
Linear Formula:
PbSO
4
CAS Number:
7446-14-2
Molecular Weight:
303.26
EC Number:
231-198-9
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Reagent Type
Core
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
307734
Lead(II) sulfate,
98%
Expand
View Pricing
254258
Lead(II) sulfate,
99.995% trace metals basis
Expand
View Pricing