Skip to Content
Merck
All Photos(3)

Documents

254258

Sigma-Aldrich

Lead(II) sulfate

99.995% trace metals basis

Synonym(s):

Anglesite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbSO4
CAS Number:
Molecular Weight:
303.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

impurities

≤55.0 ppm Trace Metal Analysis

density

6.2 g/mL at 25 °C (lit.)

SMILES string

[PbH2++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Pb/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

KEQXNNJHMWSZHK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Lead(II) sulfate uses.: Widely recognized for its role in lead-acid battery technology, Lead(II) sulfate is pivotal in the production of battery plates. It contributes to the battery′s ability to store and release energy efficiently, which is critical in automotive and renewable energy storage applications (DOI: 10.1039/d3ra04386j).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R P Purkayastha et al.
Indian journal of experimental biology, 30(12), 1184-1187 (1992-12-01)
Uptake of a few metals by V. volvacea was determined during submerged growth of the organism in sublethal concentration of each metal salt. The uptake of Pb2+ and Hg2+ was 5 and 5.23 micrograms g-1 respectively while that of Cu2+
S R Rao et al.
Waste management (New York, N.Y.), 26(5), 504-508 (2005-06-25)
Resource recovery from two metallurgical residues by flotation was investigated applying an electrostatic model to select initial conditions. The first, a sulphation roast/water leach residue, was processed to float lead sulphate, comparing dodecylamine and xanthate collectors. From the second, a
P Sanz et al.
The Journal of international medical research, 35(1), 159-163 (2007-04-06)
A 53-year-old man who had worked for 17 years manufacturing car batteries, with overt exposure to lead, developed a clinical picture initially characterized by signs of parkinsonism, followed by atypical signs such as loss of memory, reduction of eye movement
R K Sahu et al.
Molecular toxicology, 2(2), 129-136 (1989-04-01)
Sister chromatid exchanges (SCE) have been examined in human lymphocytes following in vitro treatments with metal salts, nickel sulfate, lead sulfate and sodium arsenite. All of the metal salts produced significant increases in the SCE frequencies over the levels for
Nilesh C Sharma et al.
Environmental toxicology and chemistry, 23(9), 2068-2073 (2004-09-24)
The internalized speciation of lead in roots and leaves of Sesbania drummondii, a lead hyperaccumulator, grown in lead nitrate solution was studied using x-ray absorption near-edge structure and extended x-ray absorption fine structure. Lead was predominantly present as lead acetate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service