Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Coconut oil from
Cocos nucifera
Synonym(s):
Coconut fat, Copra oil
CAS Number:
8001-31-8
EC Number:
232-282-8
Products
Product Category
lipids (2)
Brand
Sigma-Aldrich (2)
Biological Source
Melting Point (°C)
Physical Form
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
C1758
Coconut oil from
Cocos nucifera
,
low-melting solid
Expand
View Pricing
23-0310
Coconut oil from
Cocos nucifera
,
CP
Expand
View Pricing