Skip to Content
Merck
All Photos(1)

Key Documents

1711268

USP

Venlafaxine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

venlafaxine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2

InChI

1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H

InChI key

QYRYFNHXARDNFZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Venlafaxine hydrochloride USP reference standard intended for use in specified quality tests and assays.

Also used to prepare standard, system suitability, and standard stock solution for assay, impurity analysis, and performance test according to the given below monographs of United States Pharmacopeia (USP):
  • Venlafaxine Hydrochloride [1]
  • Venlafaxine Tablets [2]
  • Venlafaxine Hydrochloride Extended-Release Capsules [3]
  • Desvenlafaxine [4]

Biochem/physiol Actions

Venlafaxine is a dual serotonin-noradrenaline reuptake inhibitor that displays Ki values of 82 and 2480 nM respectively. It is a general class antidepressant.
Venlafaxine is an antidepressant. The mechanism of the antidepresant action of venlafaxine in humans is associated with its potentiation of neurotransmitter activity in the CNS. Venlafaxine is a potent inhibitor of neuronal serotonin and norepinephrine reuptake and weak inhibitor of dopamine reuptake. Venlafaxine has no significant activity for muscarinic, histaminergic, or α-1 adrenergic receptors in vitro. Venlafaxine does not possess MAO inhibitor activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Venlafaxine Hydrochloride Extended-Release Capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 4598-4598 (2022)
Venlafaxine Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(3), 4595-4595 (2017)
Desvenlafaxine
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(5), 1280-1280 (2020)
Venlafaxine Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(1), 4597-4597 (2020)
Denise G Tate et al.
Archives of physical medicine and rehabilitation, 96(2), 340-348 (2014-12-03)
To assess the role that treatment response plays in a randomized controlled trial of an antidepressant among people with spinal cord injury (SCI) diagnosed with major depressive disorder (MDD) in explaining quality of life (QOL), assessed both globally as life

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service