Skip to Content
Merck
All Photos(1)

Key Documents

1411005

USP

Methimazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Beilstein:
108646
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methimazole

manufacturer/tradename

USP

mp

144-147 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C=CNC1=S

InChI

1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)

InChI key

PMRYVIKBURPHAH-UHFFFAOYSA-N

Gene Information

human ... TPO(7173)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methimazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Methimazole Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando Bolaños et al.
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion, 54(4), 307-310 (2002-11-06)
Long-term thionamide treatment is considered one of the main resources for Graves' hyperthyroidism. Although a 54.2% remission rate in patients so treated in Mexico was previously reported, most articles have shown a wide variation over time. In the present article
K E Hill et al.
New Zealand veterinary journal, 62(4), 208-213 (2013-12-20)
To determine the pharmacokinetics of a novel lipophilic formulation of transdermal methimazole compared to oral carbimazole. Healthy cats received 5 mg carbimazole orally every 12 hours for 13 treatments (n=6), then received transdermal methimazole (n=5) at a dose of 5 mg, then
M Clementi et al.
American journal of medical genetics, 83(1), 43-46 (1999-03-17)
We report on a further case of congenital anomalies in a child exposed to methimazole during the first trimester of pregnancy (from first to seventh gestational week), and define a specific malformation pattern related to prenatal methimazole exposure and consisting
T Vogt et al.
The British journal of dermatology, 133(6), 994-996 (1995-12-01)
We report a child with scalp aplasia cutis congenita, whose mother was treated with methimazole during pregnancy. The relationship between antithyroid drug administration during pregnancy and the occurrence of scalp defects is discussed, and the pertinent literature is reviewed.
Roberto Gastaldi et al.
The Journal of pediatrics, 164(5), 1189-1194 (2014-02-13)
To evaluate clinical and biochemical features of 115 children (98 female, mean age 11.3 ± 3.5 years) with Graves disease to identify possible determinants of remission. We defined as positive outcome the improvement of clinical features and restoration of euthyroidism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service