Skip to Content
Merck
All Photos(1)

Key Documents

1134299

USP

Citalopram Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(RS)-Dimethyl(1-methyl-3,3-diphenylallyl)amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N · HCl
CAS Number:
Molecular Weight:
287.83
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

citalopram

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[N+H](C(C)C=C(c2ccccc2)c1ccccc1)(C)C

InChI

1S/C18H21N.ClH/c1-15(19(2)3)14-18(16-10-6-4-7-11-16)17-12-8-5-9-13-17;/h4-15H,1-3H3;1H

InChI key

ZVTAYVQUGPGRAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Citalopram Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Citalopram Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ebert et al.
Pharmacology & toxicology, 82(3), 157-160 (1998-04-29)
The opioid, ketobemidone, has previously been shown to be a non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist. In Denmark, ketobemidone is available in a formulation which contains ketobemidone and a spasmolytic compound, (RS)-3-dimethylamino-1,1-diphenylbut-1-ene, hydrochloride (A29), in a one to five ratio. Using
J C Raeder et al.
Acta anaesthesiologica Scandinavica, 30(7), 502-506 (1986-10-01)
A double-blind comparison of the effect of Ketogan (ketobemidone and A-29, an anticholinergic and spasmolytic agent) and morphine + scopolamine as premedication was performed in 113 women admitted for abortion and in 114 women admitted for gynaecological dilatation and curettage.
P Anderson et al.
European journal of clinical pharmacology, 19(3), 217-223 (1981-02-01)
The pharmacokinetic constants and rectal bioavailability of the narcotic analgesic ketobemidone were determined in six male patients after surgery. Plasma concentrations were measured following intravenous administration of Ketogin 2 ml, containing ketobemidone chloride 10 mg, and a spasmolytic compound N,N-dimethyl-3,3-diphenyl-1-methylallylamine
Histamine release from human basophils and isolated rat mast cells induced by ketobemidone, pethidine and the spasmolytic A29.
N Grosman
Acta pharmacologica et toxicologica, 50(1), 78-80 (1982-01-01)
M Kjaer et al.
Acta oncologica (Stockholm, Sweden), 27(5), 583-589 (1988-01-01)
Thirteen cancer patients with moderate to severe chronic pain of malignant origin on treatment with Ketogan tablets were included in an open non-randomized cross-over study comparing the analgesic effect, side effects and serum concentrations of Ketogan tablets and mixture. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service