Skip to Content
Merck
All Photos(1)

Key Documents

1087508

USP

Camphor

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Camphor, 2-Bornanone, 2-Camphanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

5.24 (vs air)

vapor pressure

4 mmHg ( 70 °C)

API family

methyl benzylidene camphor

autoignition temp.

870 °F

expl. lim.

3.5 %

manufacturer/tradename

USP

mp

178-182 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Camphor USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Camphorated Phenol Topical Gel
  • Camphorated Phenol Topical Solution
  • Eucalyptus Oil

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Siegel et al.
Pediatric clinics of North America, 33(2), 375-379 (1986-04-01)
Camphor is present in several over-the-counter compounds of questionable use and therefore may be ingested by small children. Because seizures may follow ingestion of certain amounts, appropriate treatment is needed, including the use of anticonvulsants.
Jeffrey N Love et al.
The Journal of emergency medicine, 27(1), 49-54 (2004-06-29)
Serious pediatric toxicity resulting from exposure to small amounts of camphor-containing products has long been a problem. Twenty years ago the United States Food and Drug Administration took several actions in an attempt to ameliorate this risk. Despite these changes
D E Gibson et al.
The American journal of emergency medicine, 7(1), 41-43 (1989-01-01)
Camphor ingestion is a toxic ingestion that is seen infrequently in the emergency department. It is remarkable for its rapidity of action and toxicity. A case of camphor ingestion that displayed toxic effects is presented. The pharmacology, manifestations, and management
Weiyang Chen et al.
Molecules (Basel, Switzerland), 18(5), 5434-5454 (2013-05-15)
The fragrant camphor tree (Cinnamomum camphora) and its products, such as camphor oil, have been coveted since ancient times. Having a rich history of traditional use, it was particularly used as a fumigant during the era of the Black Death
Tamera J Corte et al.
American journal of respiratory and critical care medicine, 190(2), 208-217 (2014-06-18)
Pulmonary hypertension (PH) associated with fibrotic idiopathic interstitial pneumonia (IIP; idiopathic pulmonary fibrosis and nonspecific interstitial pneumonia) confers important additional morbidity and mortality. To evaluate the safety and clinical efficacy of the dual endothelin-1 receptor antagonist bosentan in this patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service