Skip to Content
Merck
All Photos(1)

Key Documents

1078507

USP

Bupivacaine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Bupivacaine hydrochloride monohydrate, 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O · HCl · H2O
CAS Number:
Molecular Weight:
342.90
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bupivacaine

manufacturer/tradename

USP

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[N+H]1(C(CCCC1)C(=O)Nc2c(cccc2C)C)CCCC.O

InChI

1S/C18H28N2O.ClH.H2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H;1H2

InChI key

HUCIWBPMHXGLFM-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bupivacaine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Bupivacaine Hydrochloride Injection
  • Bupivacaine Hydrochloride in Dextrose Injection
  • Bupivacaine Hydrochloride and Epinephrine Injection
  • Fentanyl Citrate and Bupivacaine Hydrochloride Compounded Injection
  • Ropivacaine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bupivacaine Hydrochloride Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(3), 611-611 (2023)
Bupivacaine Hydrochloride and Epinephrine Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 612-612 (2018)
Fentanyl Citrate and Bupivacaine Hydrochloride Compounded Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(1), 1850-1850 (2020)
Bupivacaine Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(3), 610-610 (2020)
I Demirel et al.
European review for medical and pharmacological sciences, 18(7), 1067-1072 (2014-04-26)
Just as hip prosthesis, most of the patients undergoing orthopedic lower extremity surgery (OLES) belong to the advanced age group. Sciatic nerve block combined with psoas compartment block is used as a technique alternative to central neuraxial block and GA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service