Skip to Content
Merck
All Photos(1)

Key Documents

1006506

USP

Acetohydroxamic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

AHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
1739019
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acetohydroxamic acid

manufacturer/tradename

USP

mp

88-90 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(NO)=O

InChI

1S/C2H5NO2/c1-2(4)3-5/h5H,1H3,(H,3,4)

InChI key

RRUDCFGSUDOHDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetohydroxamic acid USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Acetohydroxamic Acid Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K D Lake et al.
Drug intelligence & clinical pharmacy, 19(7-8), 530-539 (1985-07-01)
Kidney stones have an overall incidence of two to three percent in western countries. In many patients, the disease process is difficult to control and recurrence rates are high: 20 to 50 percent over the subsequent ten years. The pathogenesis
Raymond L Benza et al.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation, 34(3), 329-337 (2014-07-08)
Pulmonary hypertension (PH) associated with left heart disease (World Health Organization [WHO] Group II) has previously been linked with significant morbidity and mortality. However, there are currently no approved therapies or hemodynamic monitoring systems to improve outcomes in WHO Group
Baris Gencer et al.
Atherosclerosis, 239(1), 118-124 (2015-01-15)
2013 AHA/ACC guidelines on the treatment of cholesterol advised to tailor high-intensity statin after ACS, while previous ATP-III recommended titration of statin to reach low-density lipoprotein cholesterol (LDL-C) targets. We simulated the impact of this change of paradigm on the
Saro H Armenian et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 23(6), 1109-1114 (2014-04-11)
Childhood cancer survivors are at high risk of developing congestive heart failure (CHF) compared with the general population, and there is a dose-dependent increase in CHF risk by anthracycline dose. The mechanism by which this occurs has not been fully
Heather Edgell et al.
Journal of applied physiology (Bethesda, Md. : 1985), 118(7), 839-848 (2015-01-24)
Peripheral chemoreceptor activity/sensitivity is enhanced in chronic heart failure (HF), and sensitivity is linked to greater mortality. This study aimed to determine the role of the peripheral chemoreceptor in cardiovascular control at rest and during exercise in HF patients and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service