Y0002155
Tranexamic acid impurity B
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
cis-4-(Aminomethyl)cyclohexanecarboxylic acid, cis-Tranexamic acid
About This Item
Recommended Products
Quality Level
API family
tranexamic acid
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
[N+H3]CC1CCC(CC1)C(=O)[O-]
InChI
1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)
InChI key
GYDJEQRTZSCIOI-UHFFFAOYSA-N
General description
Application
Packaging
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service