Skip to Content
Merck
All Photos(2)

Key Documents

RAD165

radiello Cartridge Adsorbents

for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Agency

EPA TO-11A
OSHA 68, 85, 1007

packaging

pk of 20

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

SS net with 2,4-DNPH coated FLORISIL®

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling Aldehydes

Looking for similar products? Visit Product Comparison Guide

General description

A radiello diffusive sampler consists of 3 parts: the diffusive body, the adsorbent tube, and the triangular support plate. For personal air sampling, a vertical adapter can be installed. The adsorbent cartridge is removed from its glass or plastic storage tube and is inserted into the diffusive body. This is then threaded onto the triangular support plate in order to place the sampler in its sampling position.
Several different Radiello cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of Radiello thermal desorption (TD) cartridge adsorbents.

Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time.

Note: To conduct air sampling using the Radiello system, Radiello diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. Radiello Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.

Application

A radiello cartridge adsorbent packed with carbograph 4 along with PerkinElmer® tube-type adsorbent packed with Tenax® TA may be used in diffusive sampling in a study, done in order to measure the sampling rate variation of four different samplers namely GABIE, 3M 3500, PerkinElmer® TA and radiello 145 diffusive samplers, while being exposed to a low concentration of volatile organic compounds (VOCs) for a long duration (1-14 days).

Legal Information

Florisil is a registered trademark of U.S. Silica Company
PerkinElmer is a registered trademark of PerkinElmer Inc.
Tenax is a registered trademark of Buchem B.V.
radiello is a trademark of Institi Clinici Scientifici Maugeri

Disclaimer

*Note: This product is not officially specified in the method, it may be suitable for use for the compound(s) listed in the method*

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Evaluation of a diffusive sampler for measurement of carbonyl compounds in air
Uchiyama, S., et al.
Atmospheric Environment, 38 (37), 6319-6326 (2004)
Volatile aldehydes in libraries and archives
Fenech, A., et al.
Atmospheric Environment, 44 (17), 2067-2073 (2010)
Ambient levels and temporal trends of VOCs, including carbonyl compounds, and ozone at Caba?eros National Park border, Spain
Villanueva, F., et al.
Atmospheric Environment, 85, 256-265 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service