Skip to Content
Merck
All Photos(1)

Key Documents

PHL83418

Zerumbone

phyproof® Reference Substance

Synonym(s):

(2E,6E,10E)-2,6,9,9-tetramethylcycloundeca-2,6,10-trien-1-one, Zerumbone (6CI,7CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O
CAS Number:
Molecular Weight:
218.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C1=C/CC(C)(C)\C=C\C(=O)\C(C)=C\CC1

InChI

1S/C15H22O/c1-12-6-5-7-13(2)14(16)9-11-15(3,4)10-8-12/h7-9,11H,5-6,10H2,1-4H3/b11-9+,12-8+,13-7+

InChI key

GIHNTRQPEMKFKO-SKTNYSRSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

TRAIL-induced apoptosis potentiator; DR4 and DR5 inducer
Zerumbone is a TRAIL-induced apoptosis potentiator. It potentiates TRAIL-induced apoptosis through the up-regulation of DR4 and DR5 expression and the down-regulation of cFLIP. Zerumbone has very little or no cytotoxic effect on the normal human endothelial cells and dermal fibroblasts. Zerumbone is a sesquiterpene isolated from in Zingiber zerumbet Smith (wild ginger).

Other Notes

This compound is commonly found in plants of the genus: curcuma zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service