Skip to Content
Merck
All Photos(1)

Documents

PHL82571

Isosilybin B

phyproof® Reference Substance

Synonym(s):

(2R,3R)-3,5,7-Trihydroxy-2-[(2S,3S)-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[1,4]dioxin-6-yl]-4-chromanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥80.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

COc1cc(ccc1O)[C@@H]2Oc3ccc(cc3O[C@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-18-7-12(3-5-16(18)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20-,23-,24-,25+/m0/s1

InChI key

FDQAOULAVFHKBX-WAABAYLZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Isosilybin B for hepatoprotective studies: Research elucidates how Isosilybin B regulates lipogenesis and fatty acid oxidation through the AMPK/SREBP-1c/PPARα pathway, offering insights into its potential as a hepatoprotective compound and its application in liver health studies (Liu et al., 2022).

  • Isosilybin B in anticancer research: The study discusses the development of a topical formulation containing silymarin complex, including Isosilybin B, highlighting its skin permeability and potential in cancer prevention through dermal application (Shankar et al., 2023).

  • Isosilybin B in antioxidant research: A study explores the antiviral efficacy of natural phytochemicals including Isosilybin B against the SARS-CoV-2 spike glycoprotein, emphasizing its role in antioxidant biochemistry and potential therapeutic applications (Aloufi et al., 2022).

Other Notes

This compound is commonly found in plants of the genus: silybum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service