Skip to Content
Merck
All Photos(1)

Key Documents

CRM06721

Supelco

Benzoylprop-ethyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17Cl2NO3
CAS Number:
Molecular Weight:
366.24
Beilstein:
2899940
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

CCOC(=O)C(C)N(C(=O)c1ccccc1)c2ccc(Cl)c(Cl)c2

InChI

1S/C18H17Cl2NO3/c1-3-24-18(23)12(2)21(14-9-10-15(19)16(20)11-14)17(22)13-7-5-4-6-8-13/h4-12H,3H2,1-2H3

InChI key

SLCGUGMPSUYJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Benzoylprop-ethyl is an obsolete post-emergence selective herbicide that inhibits lipid biosynthesis. It belongs to the class of dichloroaniline propionate compounds and is used to control the growth of wild oats and wild barley.
As per the commission regulation no 1107/2009, repealing the council directive 91/414/EEC, benzoylprop-ethyl is not approved for use in the European Union and no default MRL value has been set for the presence of benzoylprop-ethyl in products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Benzoylprop-ethyl CRM may also be used as below:

  • Simultaneous detection of 346 pesticide residues present in grapes using gas chromatography-mass spectrometry-selected ion monitoring (GC-MS-SIM) following a primary−secondary amine (PSA) matrix solid-phase dispersion (MSPD) cleanup
  • Detection and identification of 317 pesticides from fruit and vegetable samples by high-performance liquid chromatography combined with quadrupole-time-of-flight mass spectrometry (HPLCQ-TOF/MS)
  • Method development for the determination of 187 different pesticide residues in edible fungi samples using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Development of a multi-residue method for the quantification of 252 pesticides in surface water samples using liquid chromatography quadrupole-Orbitrap high-resolution tandem mass spectrometry
  • Detection and quantification of 569 pesticide residues in honeysuckle samples by ultrahigh-performance liquid chromatography (UHPLC) in combination with quadrupole-Orbitrap high-resolution mass spectrometry
  • Multi-residue analysis of 201 pesticides in 60 different medicinal herb samples using a simplified QuEChERS extraction and detection by gas chromatography-electron impact ionization triple quadrupole mass spectrometry (GC-EI-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of water deficit stress on benzoylprop-ethyl performance and physiological traits of winter wild oat (Avena sterilis subsp. ludoviciana)
Alizade S, et al.
Crop Protection, 137, 105292-105292 (2020)
Yu-Jing Lian et al.
Journal of agricultural and food chemistry, 58(17), 9428-9453 (2010-08-18)
The article demonstrates a method of simultaneous determination for 352 pesticide residues in grapes using primary-secondary amine (PSA) matrix solid phase dispersion (MSPD) cleanup and gas chromatography-mass spectrometry-selected ion monitoring (GC-MS-SIM). Grape samples (15 g) were mixed with 6 g
Determination of 187 pesticide residues in edible fungi by liquid chromatography-tandem mass spectrometry
Chang Q, et al.
Analytical Methods : Advancing Methods and Applications, 6(12), 4288- 4304 (2014)
Heng Zhou et al.
Journal of chromatography. A, 1606, 460374-460374 (2019-08-27)
A method for both qualitative screening and quantitative determination of 569 pesticides in honeysuckle was developed based on ultrahigh-performance liquid chromatography (UHPLC) coupled to quadrupole-Orbitrap high resolution mass spectrometry and an in-house executable compound database involving the theoretical masses of
Yanwei Fu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1125, 121730-121730 (2019-08-03)
Developing an analysis of multi-pesticide residues for different herbal species-ready applications is a challenge. In the present work, a comprehensive analysis was proposed for rapid detection of 201 pesticides in various medicinal herbs. Samples were extracted and cleaned up with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service