Skip to Content
Merck
All Photos(1)

Key Documents

BP815

Bis-(1,7-dihydro-6H-purine)-6,6,-disulfide

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

6,6′-Dithiodi-9H-purine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N8S2
CAS Number:
Molecular Weight:
302.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

Quality Level

API family

azathioprine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

S(Sc3ncnc4nc[nH]c43)c1ncnc2nc[nH]c21

InChI

1S/C10H6N8S2/c1-11-5-7(13-1)15-3-17-9(5)19-20-10-6-8(14-2-12-6)16-4-18-10/h1-4H,(H,11,13,15,17)(H,12,14,16,18)

InChI key

ULNKLXSTZWVSJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bis-(1,7-dihydro-6H-purine)-6,6,-disulfide BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service