95419
Elemental Impurities according to ICH Q3D Inhalation, Standard 5
TraceCERT®, in 10% HCl, applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
Agency
according to ICH Q3D
according to Ph. Eur.
according to USP
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
composition
Au, 3 mg/L
Ir, 1 mg/L
Os, 1 mg/L
Pd, 1 mg/L
Pt, 1 mg/L
Rh, 1 mg/L
Ru, 1 mg/L
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
concentration
in 10% HCl
technique(s)
ICP: suitable
format
multi-component solution
General description
Preparation Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service