Skip to Content
Merck
All Photos(1)

Documents

59167

Supelco

SUPELCOSIL LC-DP (5 µm) HPLC Columns

L × I.D. 30 cm × 4 mm, HPLC Column

Synonym(s):

SUPELCOSIL LC-DP 5UM 30CMX4.0MM HPLC COLUMN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-DP HPLC Column, 5 μm particle size, L × I.D. 30 cm × 4 mm

Agency

suitable for USP L11

Quality Level

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

6% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

30 cm × 4 mm

surface area

170 m2/g

surface coverage

surface coverage 2.4 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

phenyl phase

particle size

5 μm

pore size

120 Å pore size

pH range

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Contains a diphenyl bonded phase, which gives greater selectivity for aromatic groups compared to alkyl-type bonded phases.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

guard cartridge


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Zabrzewska et al.
Acta poloniae pharmaceutica, 71(4), 563-573 (2014-10-03)
Preservatives are chemical substances whose role is to protect medicinal products against harmful changes caused by microorganisms. They are added to sterile medicinal products, such as eye drops and multidose solutions for injections, as well as to non-sterile products, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service