Skip to Content
Merck
All Photos(1)

Documents

57017

Supelco

Supelclean LC-SAX SPE Tube

bed wt. 500 mg, volume 3 mL, pkg of 54 ea

Synonym(s):

SAX tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

material

polypropylene tube

product line

Supelclean

composition

bed wt., 500 mg

packaging

pkg of 54 ea

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix

silica gel

matrix active group

SAX phase
quaternary amine, Cl- counterion

diameter

40-45 μm

pore size

60 Å

application(s)

food and beverages

separation technique

ion exchange
ion exchange

Looking for similar products? Visit Product Comparison Guide

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Chen et al.
Ecotoxicology and environmental safety, 189, 109946-109946 (2019-11-25)
The soils near the abandoned Shimen Realgar Mine are characterized by containing extremely high contents of total and soluble arsenic. To determine the microbial reactions and environmental factors affecting the mobilization and release of arsenic from soils phase into pore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service