Skip to Content
Merck
All Photos(1)

Key Documents

567577-U

Supelco

Discovery® HS F5 (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm, Supelguard Guard Cartridge Kit with 1 Cartridge holder + 1 Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit

material

stainless steel column

Agency

suitable for USP L43

description

Supelguard Cartridge

product line

Discovery®

feature

endcapped

packaging

pkg of 1 kit

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 4 mm

surface area

300 m2/g

matrix

fully porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

5 μm

pore size

120 Å

operating pH

2-8

application(s)

food and beverages

compatibility

use to protect Discovery HS F5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Baba et al.
Journal of chromatography. A, 1354, 109-116 (2014-06-21)
Arsenic species, including arsenous acid, arsenic acid, methylarsonic acid, and dimethylarsinic acid, were determined using HPLC-ICPMS. The species were separated with a Discovery HS F5 column and a simple, volatile, and isocratic mobile phase of 0.1% (v/v) formic acid and
Ilia Brondz et al.
Journal of pharmaceutical and biomedical analysis, 43(3), 937-944 (2006-11-03)
The drug primaquine diphosphate is used for causative treatment of malaria. Using HPLC-MS and GC-MS, this research group was previously able to show that the main contaminant of primaquine is the positional isomer quinocide [I. Brondz, D. Mantzilas, U. Klein
Federica Pellati et al.
Journal of pharmaceutical and biomedical analysis, 48(2), 254-263 (2007-12-14)
In this study a pentafluorophenylpropyl (PFPP) stationary phase was applied to the fast and reliable qualitative and quantitative analysis of ephedrine alkaloids in Ephedra plant material and derivatives. A Discovery HS F5 column (150mmx4.6mm i.d., 5microm) was used, with an
Federica Pellati et al.
Journal of chromatography. A, 1165(1-2), 58-66 (2007-08-07)
In this study, the chromatographic performance of a pentafluorophenylpropyl (PFPP) stationary phase was evaluated for the rapid separation of phenethylamine alkaloids (i.e. (+/-)-octopamine, (+/-)-synephrine, tyramine, N-methyltyramine and hordenine) in Citrus aurantium plant material (fruits and peel), various Citrus species, extracts
Jennie Wang et al.
Journal of pharmaceutical and biomedical analysis, 124, 267-273 (2016-03-13)
An HPLC method for the assay of a DNA topoisomerase inhibitor, LMP776 (NSC 725776), has been developed and validated. The stress testing of LMP776 was carried out in accordance with International Conference on Harmonization (ICH) guidelines Q1A (R2) under acidic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service