Skip to Content
Merck
All Photos(1)

Key Documents

54294-U

Supelco

Carboxen® 572 for Tobacco Smoke

Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea

Synonym(s):

ORBO Rezorian Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900
NACRES:
NB.24
Pricing and availability is not currently available.

product line

Carboxen®

Quality Level

composition

Adsorbent Bed Wt:, 300 mg

packaging

pkg of 50 ea

technique(s)

active air sampling: suitable

cartridge size

3 mL

suitability

suitable for (For sampling tobacco smoke)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Rotary and linear smoke machines

Looking for similar products? Visit Product Comparison Guide

General description

Rezorian Cartridges are made of polypropylene and typically contain one bed of adsorbent with polyethylene frits. The end fittings are luer lock syringe connections that can be used to connect to sampling pump tubing,

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
ORBO is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louise Urruty et al.
Journal of agricultural and food chemistry, 50(11), 3129-3136 (2002-05-16)
To provide an efficient and running analytical tool to strawberry plant breeders who have to characterize and compare the aromatic properties of new cultivars to those already known, a HS-SPME/GC-MS analysis method has been coupled with a statistical treatment method
H C Shih et al.
Journal of occupational and environmental hygiene, 9(7), 427-436 (2012-06-02)
A solid-phase microextraction (SPME) device was used as a diffusive sampler for airborne propylene glycol ethers (PGEs), including propylene glycol monomethyl ether (PGME), propylene glycol monomethyl ether acetate (PGMEA), and dipropylene glycol monomethyl ether (DPGME). Carboxen-polydimethylsiloxane (CAR/PDMS) SPME fiber was
Noushin Rastkari et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(14-15), 1568-1574 (2009-04-28)
Carbon nanotubes (CNTs) are a kind of new carbon-based nano-materials which have drawn great attention in many application fields. The potential single-walled carbon nanotubes (SWCNTs) as solid-phase microextraction (SPME) adsorbents for the preconcentration of environmental pollutants have been investigated in
Julien Januszkiewicz et al.
Journal of chromatography. A, 1195(1-2), 16-24 (2008-05-23)
A headspace solid-phase microextraction (HS-SPME) combined with gas chromatography-mass spectrometry (GC/MS) method was developed using experimental designs to quantify the flavor of commercial Cheddar cheese and enzyme-modified Cheddar cheese (EMCC). Seven target compounds (dimethyl disulfide, hexanal, hexanol, 2-heptanone, ethyl hexanoate
Frank Sporkert et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 772(1), 45-51 (2002-05-23)
A new and in part automated headspace solid-phase microextraction method for quantitative determination of the highly toxic rodenticide fluoroacetic acid (FAA) in serum and other biological samples has been developed. FAA and deuterated acetic acid (internal standard) were extracted from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service