54225-U
Discovery® Ag-Ion SPE Tube
bed wt. 750 mg, volume 6 mL, pk of 30
Synonym(s):
Silver Ion SPE Tube
About This Item
Recommended Products
material
polypropylene tube
Quality Level
product line
Discovery®
composition
bed wt., 750 mg
packaging
pk of 30
technique(s)
solid phase extraction (SPE): suitable
surface area
480 m2/g
volume
6 mL
matrix
silica gel (irregularly shaped, acid washed)
matrix active group
benzenesulfonic acid bonding, Polymerically bonded, coated with silver ions
particle size
50 μm
pore size
0.9 mL/g pore volume
70 Å pore size
capacity
less than or equal to 1 mg (total FAMEs)
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Organic solvents, oils, and lipids
- Developed for the fractionation of FAMEs based on degree of unsaturation, and for the resolution of cis/trans isomers.
- Silver counter-ions are anchored onto an SCX support using a proprietary procedure to offer optimal resolution, performance, and capacity
- Each lot is tested and quality controlled for cis/trans FAME resolution
Footnote
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
application for SPE, suitable for GCsuitable for GC, application for SPEsuitable for GC, application for SPEOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service