52690-U
Discovery® DSC-SCX SPE Tube
bed wt. 2 g, volume 12 mL, pk of 20
Synonym(s):
Strong Cation Exchange SPE cartridge tube
About This Item
Recommended Products
material
polypropylene tube
Quality Level
composition
bed wt., 2 g
packaging
pk of 20
technique(s)
solid phase extraction (SPE): suitable
surface area
480 m2/g
volume
12 mL
matrix
Silica gel base material (irregularly shaped, acid washed)
matrix active group
sulfonic acid bonding
particle size
50 μm
pore size
0.9 mL/g pore volume
70 Å pore diameter
capacity
0.8 meq/g total capacity
application(s)
food and beverages
separation technique
ion exchange
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Organic or aqueous solutions
- A polymerically bonded, benzene sulfonic acid functional group, pKa (< 1.0)
- Counter ion is H+
- Silica support allows for use with very organic solvents (no shrinking/swelling)
- Excellent capacity (0.8 meq/g) for cleaning up solution phase combinatorial chemistry reactions (removing target molecules from reaction by-products and excess reagents)
- The presence of the benzene ring offers some mixed-mode capabilities (hydrophobic interactions) that should be considered when extracting cations from aqueous matrices
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1A - Skin Sens. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service