Skip to Content
Merck
All Photos(1)

Key Documents

48882

Supelco

ASTM® D2887 Calibration Mix

(varied), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D2887

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

concentration

(varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Related Categories

General description

ASTM® D2887 calibration mix is a mixture of C6-C44 hydrocarbons in cyclohexane.
Aroclor 1248 is a light yellow to yellow-green colored, commercially available polychlorinated biphenyl product liquid mixture, that consists of tri-, tetra-, and pentachloro congeners.

Application

It was used to calculate the retention index of compunds, during experimental study done to identify the corrosion inhibiting long-chain primary alkyl amines, using GC-MS method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Decane 11.84 % (w/w)
    Dodecane 11.78 % (w/w)
    Dotriacontane .983 % (w/w)
    Eicosane 1.966 % (w/w)
    Heptane 5.97 % (w/w)
    Hexadecane 9.8 % (w/w)
    Hexane 7.79 % (w/w)
    Hexatriacontane .983 % (w/w)
    Nonane 7.84 % (w/w)
    Octacosane .983 % (w/w)
    Octadecane 4.915 % (w/w)
    Octane 7.88 % (w/w)
    Tetracontane .983 % (w/w)
    Tetracosane 1.966 % (w/w)
    Tetradecane 11.56 % (w/w)
    Tetratetracontane .983 % (w/w)
    Undecane 11.78 % (w/w)
    See All (17)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Groundwater Chemicals Desk Reference (2007)
Identification of corrosion inhibiting long-chain primary alkyl amines by gas chromatography and gas chromatography-mass spectrometry.
Kusch, Peter, et al.
International Journal of Mass Spectrometry, 263, 45-53 (2007)
Inhibition of rat testicular androgenesis by a polychlorinated biphenyl mixture Aroclor 1248
Andric.AS, et al.
Biology of Reproduction, 62, 1882-1888 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service