Skip to Content
Merck
All Photos(1)

Key Documents

48491

Supelco

Benzo[ghi]perylene

analytical standard

Synonym(s):

1,12-Benzoperylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1913029
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

>500 °C (lit.)

mp

277-279 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56

InChI

1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H

InChI key

GYFAGKUZYNFMBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Nijegorodov et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(13), 2673-2685 (2002-01-05)
Absorption. fluorescence and laser properties of perylene, benzo(ghi)perylene and coronene are studied experimentally (under the same conditions) and quantum chemically at room (293 K) and at low (77 and 4 K) temperatures and direct comparison is made between the results
Stepan Boitsov et al.
Marine environmental research, 68(5), 236-245 (2009-08-01)
Polycyclic aromatic hydrocarbons (PAH) were measured in sediment cores from 13 locations in South-Western Barents Sea as part of a detailed study of the Norwegian seabed under the MAREANO program. The generally low PAH levels found, an average around 200
S H Cherng et al.
Toxicology and applied pharmacology, 170(1), 63-68 (2001-01-06)
Although benzo[g,h,i]perylene (BghiP) has been found to promote the carcinogenesis of benzo[a]pyrene (BaP) in animal models, not much is known about this cocarcinogenic mechanism. In this study, human hepatoma HepG2 cells cotreated with BaP and BghiP were used as a
Nobuyasu Itoh et al.
Analytica chimica acta, 615(1), 47-53 (2008-04-29)
Microwave-assisted extraction using 1M KOH/methanol (alkaline-MAE) in combination with solid-phase extraction treatment was developed and applied to polycyclic aromatic hydrocarbons (PAHs) in a sediment sample. Although various conditions were examined (100 or 150 degrees C for 10 or 30 min)
Py and BPe solvent polarity scales: effect of temperature on pyrene and benzo[ghi]perylene fluorescence spectra.
R Waris et al.
The Analyst, 113(9), 1465-1467 (1988-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service