Skip to Content
Merck
All Photos(1)

Documents

47794

Supelco

1,2,3-Trichloropropane

analytical standard

Synonym(s):

Glycerol trichlorohydrin, Trichlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2ClCHClCH2Cl
CAS Number:
Molecular Weight:
147.43
Beilstein:
1732068
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.484 (lit.)

bp

156 °C (lit.)

mp

−14 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

ClCC(Cl)CCl

InChI

1S/C3H5Cl3/c4-1-3(6)2-5/h3H,1-2H2

InChI key

CFXQEHVMCRXUSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Kidney,Liver,Mucous membranes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

165.2 °F - DIN 51758

Flash Point(C)

74 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ghufrana Samin et al.
Environmental science and pollution research international, 19(8), 3067-3078 (2012-08-10)
1,2,3-Trichloropropane (TCP) is a persistent groundwater pollutant and a suspected human carcinogen. It is also is an industrial chemical waste that has been formed in large amounts during epichlorohydrin manufacture. In view of the spread of TCP via groundwater and
Eakalak Khan et al.
Journal of hazardous materials, 161(2-3), 1024-1034 (2008-05-27)
The mineralization and biodegradability increase and their combination of two traditional and two relatively new organic contaminants by Fenton reagents with three different types of iron, Fe(2+), Fe(3+), and Fe(0) were investigated. The traditional contaminants examined were trichloroethene (TCE) and
Vaishnavi Sarathy et al.
Environmental science & technology, 44(2), 787-793 (2009-12-17)
1,2,3-Trichloropropane (TCP) is an emerging contaminant because of increased recognition of its occurrence in groundwater, potential carcinogenicity, and resistance to natural attenuation. The physical and chemical properties of TCP make it difficult to remediate, with all conventional options being relatively
Marcin Podsiadło et al.
Acta crystallographica. Section B, Structural science, 62(Pt 6), 1071-1077 (2006-11-17)
The structure of 1,2,3-trichloropropane, ClCH2CHClCH2Cl, in-situ crystallized in a diamond-anvil cell, has been determined by single-crystal X-ray diffraction at 0.28 and 0.35 GPa. A melting point at 295 K and 0.22 GPa has been determined. The molecular conformation of aliphatic
Jan G E van Leeuwen et al.
Chembiochem : a European journal of chemical biology, 13(1), 137-148 (2011-11-24)
We used directed evolution to obtain enantiocomplementary haloalkane dehalogenase variants that convert the toxic waste compound 1,2,3-trichloropropane (TCP) into highly enantioenriched (R)- or (S)-2,3-dichloropropan-1-ol, which can easily be converted into optically active epichlorohydrins-attractive intermediates for the synthesis of enantiopure fine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service